Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTTCCCGGGTGAGTGTCCAGGCA[C/T]GCCAGCGGAACGGCCCGCGGGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603946 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELLS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HELLS - helicase, lymphoid-specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289067.1 | 157 | Missense Mutation | ACG,ATG | T,M 1 | NP_001275996.1 | |
NM_001289068.1 | 157 | Intron | NP_001275997.1 | |||
NM_001289069.1 | 157 | Missense Mutation | ACG,ATG | T,M 1 | NP_001275998.1 | |
NM_001289070.1 | 157 | Missense Mutation | ACG,ATG | T,M 1 | NP_001275999.1 | |
NM_001289071.1 | 157 | UTR 5 | NP_001276000.1 | |||
NM_001289072.1 | 157 | Missense Mutation | ACG,ATG | T,M 1 | NP_001276001.1 | |
NM_001289073.1 | 157 | UTR 5 | NP_001276002.1 | |||
NM_001289074.1 | 157 | UTR 5 | NP_001276003.1 | |||
NM_001289075.1 | 157 | UTR 5 | NP_001276004.1 | |||
NM_018063.4 | 157 | Missense Mutation | ACG,ATG | T,M 1 | NP_060533.2 |
TBC1D12 - TBC1 domain family member 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |