Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTAAGAAAGCTAAGAAGAAGGC[C/G]TCATCCAGTGACAGTGAGGACAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOLC1 - nucleolar and coiled-body phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284388.1 | 481 | Silent Mutation | GCC,GCG | A,A 82 | NP_001271317.1 | |
NM_001284389.1 | 481 | Silent Mutation | GCC,GCG | A,A 83 | NP_001271318.1 | |
NM_004741.4 | 481 | Silent Mutation | GCC,GCG | A,A 82 | NP_004732.2 | |
XM_005270273.1 | 481 | Silent Mutation | GCC,GCG | A,A 83 | XP_005270330.1 |
PPRC1 - peroxisome proliferator-activated receptor gamma, coactivator-related 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |