Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGGAACGTTCATGGTCAGCTG[G/T]GTGGTGTAGCTGCGGTAAAAGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A28 - solute carrier family 25 member 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031212.3 | 705 | Silent Mutation | ACA,ACC | T,T 230 | NP_112489.3 | |
XM_005270208.1 | 705 | Silent Mutation | ACA,ACC | T,T 131 | XP_005270265.1 | |
XM_005270211.4 | 705 | Silent Mutation | ACA,ACC | T,T 131 | XP_005270268.1 | |
XM_006718005.3 | 705 | Silent Mutation | ACA,ACC | T,T 131 | XP_006718068.1 | |
XM_006718006.3 | 705 | Intron | XP_006718069.1 | |||
XM_011540239.2 | 705 | Silent Mutation | ACA,ACC | T,T 140 | XP_011538541.1 | |
XM_011540240.1 | 705 | Silent Mutation | ACA,ACC | T,T 131 | XP_011538542.1 | |
XM_011540244.2 | 705 | Intron | XP_011538546.1 | |||
XM_017016741.1 | 705 | Silent Mutation | ACA,ACC | T,T 131 | XP_016872230.1 | |
XM_017016742.1 | 705 | Silent Mutation | ACA,ACC | T,T 75 | XP_016872231.1 |