Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTAGACCAGCAGCTTATGAGC[A/G]ATGATGAGGACAGCCTTCAGGATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610258 MIM: 601396 | ||||||||||||||||||||
Literature Links: |
SEC31B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC31B - SEC31 homolog B, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015490.3 | 3621 | Silent Mutation | ATC,ATT | I,I 1173 | NP_056305.1 |
WNT8B - Wnt family member 8B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |