Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCCCGGCAGGAAGCGCGCCCCC[C/G]GGGCCGGTCCCGGAGGGCTCGATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605482 | ||||||||||||||||||||
Literature Links: |
GSTO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTO1 - glutathione S-transferase omega 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191002.1 | 267 | Silent Mutation | CCC,CCG | P,P 16 | NP_001177931.1 | |
NM_001191003.1 | 267 | UTR 5 | NP_001177932.1 | |||
NM_004832.2 | 267 | Silent Mutation | CCC,CCG | P,P 16 | NP_004823.1 |