Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCAGTGATCCTTCTAAGGGGGA[C/T]GTAGCTGGTTGATGAAATGCTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFMBT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFMBT2 - Scm-like with four mbt domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018039.1 | 4050 | UTR 3 | NP_001018049.1 | |||
NM_001029880.2 | 4050 | UTR 3 | NP_001025051.1 | |||
XM_005252551.2 | 4050 | UTR 3 | XP_005252608.1 | |||
XM_006717490.2 | 4050 | UTR 3 | XP_006717553.1 | |||
XM_011519611.1 | 4050 | UTR 3 | XP_011517913.1 | |||
XM_017016466.1 | 4050 | UTR 3 | XP_016871955.1 |