Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTATGAATATATTGATGAGATGT[C/T]TAGCATGTGAAAAATTACATTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606808 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929073 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929073 - uncharacterized LOC101929073 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYO3A - myosin IIIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017433.4 | Intron | NP_059129.3 | ||||
XM_011519498.2 | Intron | XP_011517800.1 | ||||
XM_011519499.1 | Intron | XP_011517801.1 | ||||
XM_011519500.2 | Intron | XP_011517802.1 | ||||
XM_011519502.1 | Intron | XP_011517804.1 | ||||
XM_011519503.1 | Intron | XP_011517805.1 | ||||
XM_011519504.1 | Intron | XP_011517806.1 | ||||
XM_011519505.1 | Intron | XP_011517807.1 | ||||
XM_011519506.2 | Intron | XP_011517808.1 | ||||
XM_011519508.2 | Intron | XP_011517810.1 | ||||
XM_011519510.1 | Intron | XP_011517812.1 | ||||
XM_011519511.2 | Intron | XP_011517813.1 | ||||
XM_011519512.1 | Intron | XP_011517814.1 | ||||
XM_011519513.2 | Intron | XP_011517815.1 |