Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACCAGGTCATCAGGGCTTTGGG[C/T]GTTGGCCTGGGAGCTGGCTGGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC20 - leucine rich repeat containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278211.1 | 1661 | UTR 3 | NP_001265140.1 | |||
NM_001278212.1 | 1661 | UTR 3 | NP_001265141.1 | |||
NM_001278213.1 | 1661 | UTR 3 | NP_001265142.1 | |||
NM_001278214.1 | 1661 | UTR 3 | NP_001265143.1 | |||
NM_018205.3 | 1661 | UTR 3 | NP_060675.1 | |||
NM_018239.3 | 1661 | UTR 3 | NP_060709.2 | |||
NM_207119.2 | 1661 | UTR 3 | NP_997002.1 | |||
XM_017016374.1 | 1661 | UTR 3 | XP_016871863.1 | |||
XM_017016375.1 | 1661 | UTR 3 | XP_016871864.1 |