Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTTTACCTAATAAGCTATCTTC[C/T]TTACTTTAGCTGTAGAAGGCTGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606615 MIM: 602140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIF1AN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIF1AN - hypoxia inducible factor 1 alpha subunit inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017902.2 | Intron | NP_060372.2 | ||||
XM_011539940.2 | Intron | XP_011538242.1 | ||||
XM_011539941.1 | Intron | XP_011538243.1 |
NDUFB8 - NADH:ubiquinone oxidoreductase subunit B8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |