Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCATGGCCCCTAAGTGGCCATT[A/G]ATGACATGGCATAAGATCACAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602714 MIM: 611640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM12 - ADAM metallopeptidase domain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288973.1 | 5255 | UTR 3 | NP_001275902.1 | |||
NM_001288974.1 | 5255 | Intron | NP_001275903.1 | |||
NM_001288975.1 | 5255 | Intron | NP_001275904.1 | |||
NM_003474.5 | 5255 | UTR 3 | NP_003465.3 | |||
NM_021641.4 | 5255 | Intron | NP_067673.2 | |||
XM_017016705.1 | 5255 | UTR 3 | XP_016872194.1 | |||
XM_017016706.1 | 5255 | UTR 3 | XP_016872195.1 |
FANK1 - fibronectin type III and ankyrin repeat domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |