Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAATTACCAGAGGCCGCCGACTC[A/G]TTTGGGGAAATTGCTTCTGGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612302 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRA1 - adhesion G protein-coupled receptor A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083909.2 | 507 | Intron | NP_001077378.1 | |||
NM_001291085.1 | 507 | Intron | NP_001278014.1 | |||
XM_005252695.1 | 507 | Silent Mutation | TCA,TCG | S,S 169 | XP_005252752.1 | |
XM_011540273.1 | 507 | Intron | XP_011538575.1 | |||
XM_017016779.1 | 507 | Intron | XP_016872268.1 |