Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTCCCTTACGATCATGGATGAC[A/G]CTGTGAGAACCCTATCATGGCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKI67 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKI67 - marker of proliferation Ki-67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145966.1 | 9217 | UTR 3 | NP_001139438.1 | |||
NM_002417.4 | 9217 | UTR 3 | NP_002408.3 | |||
XM_006717864.3 | 9217 | UTR 3 | XP_006717927.2 | |||
XM_011539818.2 | 9217 | UTR 3 | XP_011538120.1 |