Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGATCTTTGCCCCTCTATAAAA[C/T]GCCACTTCGCAAGGTTTTGAGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611932 MIM: 609851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CISD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CISD1 - CDGSH iron sulfur domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018464.4 | Intron | NP_060934.1 | ||||
XM_017016409.1 | Intron | XP_016871898.1 |
IPMK - inositol polyphosphate multikinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152230.4 | Intron | NP_689416.1 | ||||
XM_011539565.2 | Intron | XP_011537867.1 | ||||
XM_017016013.1 | Intron | XP_016871502.1 |