Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAATGAAGTGACATTATTCTTTG[C/T]AGCTGGTTACTTACATTTCAAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612189 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PBLD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PBLD - phenazine biosynthesis like protein domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033083.1 | 1583 | Intron | NP_001028255.1 | |||
NM_022129.3 | 1583 | UTR 3 | NP_071412.2 | |||
XM_005270028.3 | 1583 | UTR 3 | XP_005270085.1 | |||
XM_011540060.2 | 1583 | UTR 3 | XP_011538362.1 | |||
XM_017016513.1 | 1583 | UTR 3 | XP_016872002.1 | |||
XM_017016514.1 | 1583 | Intron | XP_016872003.1 |