Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTACCAGAGTTTTAATATATAA[C/T]TTCTTGATTTAGCTTTGGGCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGPL1 - sphingosine-1-phosphate lyase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003901.3 | Intron | NP_003892.2 | ||||
XM_005270263.1 | Intron | XP_005270320.1 | ||||
XM_006718053.1 | Intron | XP_006718116.1 | ||||
XM_011540316.2 | Intron | XP_011538618.1 | ||||
XM_011540317.1 | Intron | XP_011538619.1 | ||||
XM_011540318.1 | Intron | XP_011538620.1 |