Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCATTTTTAGCCCTAGCAGGCA[G/T]GAAGTGTGGCCATGCTCTCTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600926 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRE - protein tyrosine phosphatase, receptor type E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316676.1 | Intron | NP_001303605.1 | ||||
NM_001316677.1 | Intron | NP_001303606.1 | ||||
NM_001323354.1 | Intron | NP_001310283.1 | ||||
NM_001323355.1 | Intron | NP_001310284.1 | ||||
NM_001323356.1 | Intron | NP_001310285.1 | ||||
NM_001323357.1 | Intron | NP_001310286.1 | ||||
NM_006504.5 | Intron | NP_006495.1 | ||||
NM_130435.4 | Intron | NP_569119.1 | ||||
XM_005252691.2 | Intron | XP_005252748.1 | ||||
XM_011539994.2 | Intron | XP_011538296.1 | ||||
XM_011539995.1 | Intron | XP_011538297.1 | ||||
XM_011539996.1 | Intron | XP_011538298.1 | ||||
XM_011539998.1 | Intron | XP_011538300.1 | ||||
XM_017016467.1 | Intron | XP_016871956.1 | ||||
XM_017016468.1 | Intron | XP_016871957.1 | ||||
XM_017016469.1 | Intron | XP_016871958.1 |