Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGGCAGCAGGGCACGGCCAAG[A/G]TAACTGTAGATTGGAGATTGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603451 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDB1 - LIM domain binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113407.2 | Intron | NP_001106878.1 | ||||
NM_001321612.1 | Intron | NP_001308541.1 | ||||
NM_003893.4 | Intron | NP_003884.1 | ||||
XM_017016867.1 | Intron | XP_016872356.1 | ||||
XM_017016868.1 | Intron | XP_016872357.1 | ||||
XM_017016869.1 | Intron | XP_016872358.1 |