Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTTGTGGACGATCAGCCTGTACT[G/T]CCTGGCCAGGCACAGCTCCTCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609044 MIM: 180250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FFAR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FFAR4 - free fatty acid receptor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBP4 - retinol binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323517.1 | 628 | Missense Mutation | AAG,CAG | K,Q 182 | NP_001310446.1 | |
NM_001323518.1 | 628 | Missense Mutation | AAG,CAG | K,Q 180 | NP_001310447.1 | |
NM_006744.3 | 628 | Missense Mutation | AAG,CAG | K,Q 182 | NP_006735.2 |