Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATACAACTGAGCCCAAAATGCT[C/T]CACTGCAATAGTAATGGTCTGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTER - phosphotriesterase related | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001484.2 | Intron | NP_001001484.1 | ||||
NM_001261836.1 | Intron | NP_001248765.1 | ||||
NM_001261837.1 | Intron | NP_001248766.1 | ||||
NM_001261838.1 | Intron | NP_001248767.1 | ||||
NM_030664.4 | Intron | NP_109589.2 | ||||
XM_017016927.1 | Intron | XP_016872416.1 | ||||
XM_017016928.1 | Intron | XP_016872417.1 | ||||
XM_017016929.1 | Intron | XP_016872418.1 | ||||
XM_017016930.1 | Intron | XP_016872419.1 |