Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAAACAGTAGATGAAGCAAGT[G/T]TGGCAAAATCTTGATAACTGTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614656 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PALD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PALD1 - phosphatase domain containing, paladin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014431.2 | Intron | NP_055246.2 | ||||
XM_011539637.2 | Intron | XP_011537939.1 | ||||
XM_011539638.2 | Intron | XP_011537940.1 | ||||
XM_017016072.1 | Intron | XP_016871561.1 |