Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTCAGCCCCAGATAGGGTTGCA[C/T]CTTGGAGTGTCCTCATTTTATTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN14 - tetraspanin 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128309.1 | Intron | NP_001121781.1 | ||||
NM_030927.2 | Intron | NP_112189.2 | ||||
XM_006717992.3 | Intron | XP_006718055.1 | ||||
XM_011540219.2 | Intron | XP_011538521.1 | ||||
XM_011540220.2 | Intron | XP_011538522.1 | ||||
XM_011540221.2 | Intron | XP_011538523.1 | ||||
XM_011540222.2 | Intron | XP_011538524.1 | ||||
XM_017016707.1 | Intron | XP_016872196.1 | ||||
XM_017016708.1 | Intron | XP_016872197.1 | ||||
XM_017016709.1 | Intron | XP_016872198.1 | ||||
XM_017016710.1 | Intron | XP_016872199.1 | ||||
XM_017016711.1 | Intron | XP_016872200.1 | ||||
XM_017016712.1 | Intron | XP_016872201.1 | ||||
XM_017016713.1 | Intron | XP_016872202.1 |