Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAACATTGACCCCAAACCATAG[C/T]AAGTGCATTAAGGAGGGGAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBL2 - mannose binding lectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000242.2 | 3748 | Intron | NP_000233.1 | |||
XM_006717861.3 | 3748 | UTR 3 | XP_006717924.1 | |||
XM_011539816.2 | 3748 | UTR 3 | XP_011538118.1 |