Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTGATCGTACACTTAAGGAATC[C/T]GAATTGTAGCAGCACCACAAAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G12B - phospholipase A2 group XIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318124.1 | Intron | NP_001305053.1 | ||||
NM_001318125.1 | Intron | NP_001305054.1 | ||||
NM_032562.4 | Intron | NP_115951.2 | ||||
XM_005270244.4 | Intron | XP_005270301.1 |