Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGATGTTGTCTCTGACTCTGA[A/G]GTAAGTGTTATTTGTCGCTTAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDRF1 - erythroid differentiation regulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202438.1 | 677 | Silent Mutation | GAA,GAG | E,E 128 | NP_001189367.1 | |
NM_015608.2 | 677 | Silent Mutation | GAA,GAG | E,E 128 | NP_056423.2 | |
XM_011539622.1 | 677 | Intron | XP_011537924.1 | |||
XM_017016066.1 | 677 | UTR 5 | XP_016871555.1 |
FLJ37035 - uncharacterized LOC399821 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |