Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATTCCTTATTGGAGGACAGACA[A/G]TAATAAAAATCTAGGTAATATTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613921 MIM: 609360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPJ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPJ - lipase family member J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010939.2 | 74 | Missense Mutation | AAT,AGT | N,S 39 | NP_001010939.2 | |
XM_005269542.3 | 74 | UTR 5 | XP_005269599.1 | |||
XM_006717635.2 | 74 | Missense Mutation | AAT,AGT | N,S 39 | XP_006717698.1 | |
XM_011539314.2 | 74 | Missense Mutation | AAT,AGT | N,S 39 | XP_011537616.1 | |
XM_011539315.2 | 74 | Missense Mutation | AAT,AGT | N,S 39 | XP_011537617.1 | |
XM_011539317.2 | 74 | Missense Mutation | AAT,AGT | N,S 39 | XP_011537619.1 | |
XM_011539318.2 | 74 | Intron | XP_011537620.1 | |||
XM_011539319.1 | 74 | UTR 5 | XP_011537621.1 | |||
XM_011539320.2 | 74 | Intron | XP_011537622.1 | |||
XM_017015740.1 | 74 | Missense Mutation | AAT,AGT | N,S 39 | XP_016871229.1 |
RNLS - renalase, FAD dependent amine oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |