Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCTTGGGCCCTCTTGGCTTCC[A/G]TGTGTCCTCTTTCCAGGCATGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929574 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929574 - uncharacterized LOC101929574 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2D4B - SH2 domain containing 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145719.1 | Intron | NP_001139191.1 | ||||
NM_207372.2 | Intron | NP_997255.2 | ||||
XM_017016236.1 | Intron | XP_016871725.1 |