Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGGTCTGTTCCGCCTCCGTTTCG[A/G]GGTCCACCCGAAAAGCTTTCAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614142 MIM: 610287 MIM: 602327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUEDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUEDC2 - CUE domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXL15 - F-box and leucine rich repeat protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024326.3 | 1166 | UTR 5 | NP_077302.3 | |||
XM_005270149.2 | 1166 | Intron | XP_005270206.1 | |||
XM_005270150.4 | 1166 | Intron | XP_005270207.1 | |||
XM_005270151.3 | 1166 | Intron | XP_005270208.1 | |||
XM_017016631.1 | 1166 | Intron | XP_016872120.1 |
PSD - pleckstrin and Sec7 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270965.1 | 1166 | Intron | NP_001257894.1 | |||
NM_001270966.1 | 1166 | Intron | NP_001257895.1 | |||
NM_002779.4 | 1166 | Intron | NP_002770.3 | |||
XM_011539968.1 | 1166 | Intron | XP_011538270.1 | |||
XM_011539969.2 | 1166 | Intron | XP_011538271.1 | |||
XM_017016433.1 | 1166 | Intron | XP_016871922.1 | |||
XM_017016434.1 | 1166 | Intron | XP_016871923.1 |