Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCAAAGTATTACAGTAAATTCT[C/T]ATTTATTCTGAATCCAAACTACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTKN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RTKN2 - rhotekin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282941.1 | Intron | NP_001269870.1 | ||||
NM_145307.3 | Intron | NP_660350.2 | ||||
XM_011539456.2 | Intron | XP_011537758.1 | ||||
XM_011539457.2 | Intron | XP_011537759.1 | ||||
XM_011539460.2 | Intron | XP_011537762.1 | ||||
XM_017015842.1 | Intron | XP_016871331.1 | ||||
XM_017015843.1 | Intron | XP_016871332.1 | ||||
XM_017015844.1 | Intron | XP_016871333.1 |