Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGATAGATAAAATTTAAAAAGAA[A/G]AGCAAAAAGACATACTGGGAGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf67 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf67 - chromosome 10 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153714.2 | Intron | NP_714925.2 | ||||
XM_011519440.1 | Intron | XP_011517742.1 | ||||
XM_011519441.1 | Intron | XP_011517743.1 | ||||
XM_011519442.2 | Intron | XP_011517744.1 | ||||
XM_011519443.1 | Intron | XP_011517745.1 | ||||
XM_011519444.2 | Intron | XP_011517746.1 | ||||
XM_011519445.2 | Intron | XP_011517747.1 | ||||
XM_011519446.1 | Intron | XP_011517748.1 | ||||
XM_017016029.1 | Intron | XP_016871518.1 | ||||
XM_017016030.1 | Intron | XP_016871519.1 | ||||
XM_017016031.1 | Intron | XP_016871520.1 |