Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGACATTCTCTAATCCTAGGAGG[A/G]AGATATCATTAAATCACTGGTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604695 MIM: 615570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL3 - ADP ribosylation factor like GTPase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFXN2 - sideroflexin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178858.4 | 21 | Intron | NP_849189.1 | |||
XM_011539261.2 | 21 | Intron | XP_011537563.1 | |||
XM_011539262.2 | 21 | Intron | XP_011537564.1 | |||
XM_011539263.2 | 21 | Intron | XP_011537565.1 | |||
XM_011539264.2 | 21 | Intron | XP_011537566.1 | |||
XM_017015663.1 | 21 | UTR 5 | XP_016871152.1 |