Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAAGGAGGTACTTGTTAAATCT[A/G]TGTGTTTTTATTATAGACATCGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156569 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGMT - O-6-methylguanine-DNA methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002412.4 | Intron | NP_002403.2 | ||||
XM_005252682.2 | Intron | XP_005252739.1 | ||||
XM_017016275.1 | Intron | XP_016871764.1 |