Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATTAGCTGTCTGGGGAAGCTAA[C/T]TTCCTACAGTGGGTTTTAGTTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP19 - Rho GTPase activating protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204300.1 | 4967 | UTR 3 | NP_001191229.1 | |||
NM_001256423.1 | 4967 | UTR 3 | NP_001243352.1 | |||
NM_032900.5 | 4967 | UTR 3 | NP_116289.4 |
ARHGAP19-SLIT1 - ARHGAP19-SLIT1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |