Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAAATATTCTATCGGATCCACT[C/G]GGCGAACTCTTGCCACTTCTGCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615154 | ||||||||||||||||||||
Literature Links: |
DYDC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYDC1 - DPY30 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001269053.1 | 162 | Missense Mutation | CCA,CGA | P,R 26 | NP_001255982.1 | |
NM_138812.3 | 162 | Missense Mutation | CCA,CGA | P,R 26 | NP_620167.1 | |
XM_005269549.4 | 162 | Missense Mutation | CCA,CGA | P,R 26 | XP_005269606.1 | |
XM_005269550.4 | 162 | Missense Mutation | CCA,CGA | P,R 26 | XP_005269607.1 | |
XM_011539334.2 | 162 | Missense Mutation | CCA,CGA | P,R 26 | XP_011537636.1 | |
XM_011539335.1 | 162 | Missense Mutation | CCA,CGA | P,R 26 | XP_011537637.1 | |
XM_017015749.1 | 162 | Missense Mutation | CCA,CGA | P,R 26 | XP_016871238.1 |
DYDC2 - DPY30 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270041.1 | 162 | Intron | NP_001256970.1 | |||
NM_001270042.1 | 162 | Intron | NP_001256971.1 | |||
NM_032372.5 | 162 | Intron | NP_115748.1 | |||
XM_011540267.2 | 162 | Intron | XP_011538569.1 | |||
XM_011540268.1 | 162 | Intron | XP_011538570.1 | |||
XM_011540270.2 | 162 | Intron | XP_011538572.1 |