Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGGTACTAGAACTGATCACCC[A/G]TGAAGTGCACGTGGGTGTGTACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616616 MIM: 614531 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSGALNACT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSGALNACT2 - chondroitin sulfate N-acetylgalactosaminyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGEF1A - RasGEF domain family member 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282862.1 | 3045 | UTR 3 | NP_001269791.1 | |||
NM_145313.3 | 3045 | UTR 3 | NP_660356.2 | |||
XM_005271809.3 | 3045 | Intron | XP_005271866.1 | |||
XM_011539500.2 | 3045 | Intron | XP_011537802.1 |