Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAGGCATGTTTTCCTCTGGCAG[C/G]GTCAATCATGCTTAGCTGCAGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LARP4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LARP4B - La ribonucleoprotein domain family member 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015155.2 | 4511 | UTR 3 | NP_055970.1 | |||
XM_005252432.3 | 4511 | Intron | XP_005252489.1 | |||
XM_005252435.3 | 4511 | Intron | XP_005252492.1 | |||
XM_011519435.2 | 4511 | Intron | XP_011517737.1 | |||
XM_017015987.1 | 4511 | UTR 3 | XP_016871476.1 | |||
XM_017015988.1 | 4511 | UTR 3 | XP_016871477.1 | |||
XM_017015989.1 | 4511 | UTR 3 | XP_016871478.1 | |||
XM_017015990.1 | 4511 | Intron | XP_016871479.1 | |||
XM_017015991.1 | 4511 | Intron | XP_016871480.1 | |||
XM_017015992.1 | 4511 | UTR 3 | XP_016871481.1 | |||
XM_017015993.1 | 4511 | UTR 3 | XP_016871482.1 | |||
XM_017015994.1 | 4511 | UTR 3 | XP_016871483.1 | |||
XM_017015995.1 | 4511 | Intron | XP_016871484.1 |