Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGAAGGGGCCAGCAAAAACAAA[A/T]AAGTTGAAAATCGAAAAGAGAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612333 MIM: 615204 | ||||||||||||||||||||
Literature Links: |
MIR1307 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR1307 - microRNA 1307 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDCD11 - programmed cell death 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014976.1 | 292 | Missense Mutation | AAA,AAT | K,N 55 | NP_055791.1 | |
XM_005269647.3 | 292 | Missense Mutation | AAA,AAT | K,N 55 | XP_005269704.1 | |
XM_011539538.2 | 292 | Missense Mutation | AAA,AAT | K,N 55 | XP_011537840.1 | |
XM_011539539.2 | 292 | Missense Mutation | AAA,AAT | K,N 55 | XP_011537841.1 | |
XM_011539540.1 | 292 | UTR 5 | XP_011537842.1 |
USMG5 - up-regulated during skeletal muscle growth 5 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |