Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTAGGAAGTCAAGGAAGGTCTCA[C/T]GGAGAGGGAGTCAAACTGAAACTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605506 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS26A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS26A - VPS26, retromer complex component A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035260.2 | Intron | NP_001030337.1 | ||||
NM_001318944.1 | Intron | NP_001305873.1 | ||||
NM_001318945.1 | Intron | NP_001305874.1 | ||||
NM_001318946.1 | Intron | NP_001305875.1 | ||||
NM_004896.4 | Intron | NP_004887.2 |