Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCGGCAGCACCAACGGAAACATG[A/G]CTCTGTTCAGCCTCCCAGAGGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDHR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDHR1 - cadherin related family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171971.2 | 244 | Missense Mutation | ACT,GCT | T,A 40 | NP_001165442.1 | |
NM_033100.3 | 244 | Missense Mutation | ACT,GCT | T,A 40 | NP_149091.1 | |
XM_011540337.1 | 244 | Missense Mutation | ACT,GCT | T,A 98 | XP_011538639.1 | |
XM_011540338.1 | 244 | Missense Mutation | ACT,GCT | T,A 98 | XP_011538640.1 | |
XM_011540339.1 | 244 | UTR 5 | XP_011538641.1 | |||
XM_011540340.2 | 244 | Missense Mutation | ACT,GCT | T,A 98 | XP_011538642.1 |