Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAAGAAGCTGCTGAAAACAGAA[A/G]TTCACCTGCACATCTTGTTTTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HECTD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HECTD2 - HECT domain E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284274.1 | 384 | Missense Mutation | AAT,AGT | N,S 77 | NP_001271203.1 | |
NM_173497.3 | 384 | Missense Mutation | AAT,AGT | N,S 77 | NP_775768.3 | |
NM_182765.4 | 384 | Missense Mutation | AAT,AGT | N,S 77 | NP_877497.3 | |
XM_017015750.1 | 384 | UTR 5 | XP_016871239.1 | |||
XM_017015751.1 | 384 | Missense Mutation | AAT,AGT | N,S 77 | XP_016871240.1 | |
XM_017015752.1 | 384 | Intron | XP_016871241.1 |
HECTD2-AS1 - HECTD2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |