Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGGATGAAGAAGGAGCATGTG[C/T]TTCACTGCCAGTTCTCCGCGTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC123 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC123 - cell division cycle 123 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006023.2 | 120 | Missense Mutation | CTT,TTT | L,F 7 | NP_006014.2 | |
XM_005252638.3 | 120 | Missense Mutation | CTT,TTT | L,F 7 | XP_005252695.1 |
NUDT5 - nudix hydrolase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321647.1 | 120 | Intron | NP_001308576.1 | |||
NM_001321648.1 | 120 | Intron | NP_001308577.1 | |||
NM_014142.3 | 120 | Intron | NP_054861.2 |