Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCCCCACTCACCTGCTACCTG[G/T]TACTCTGTTGTAAATATGGACGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147730 | ||||||||||||||||||||
Literature Links: |
IL2RA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL2RA - interleukin 2 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000417.2 | 936 | Nonsense Mutation | TAA,TAC | *,Y 239 | NP_000408.1 | |
NM_001308242.1 | 936 | Nonsense Mutation | TAA,TAC | *,Y 167 | NP_001295171.1 | |
NM_001308243.1 | 936 | Nonsense Mutation | TAA,TAC | *,Y 143 | NP_001295172.1 |