Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACAGTCACCATGGCCACCCAA[A/G]CCACCAGAGCCACAGCCTGCCTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610734 MIM: 613597 | ||||||||||||||||||||
Literature Links: |
ANKRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD2 - ankyrin repeat domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C10orf62 - chromosome 10 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009997.2 | 840 | Missense Mutation | AAC,AGC | N,S 212 | NP_001009997.2 |
HOGA1 - 4-hydroxy-2-oxoglutarate aldolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134670.1 | 840 | Intron | NP_001128142.1 | |||
NM_138413.3 | 840 | Intron | NP_612422.2 |