Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCGGGGGGCCAGCTCCTCCCC[A/G]CCACCACGGCCCTCCTGGGCCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606075 MIM: 610454 MIM: 612971 | ||||||||||||||||||||
Literature Links: |
C10orf2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C10orf2 - chromosome 10 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LZTS2 - leucine zipper tumor suppressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318099.1 | 349 | Missense Mutation | CAC,CGC | H,R 46 | NP_001305028.1 | |
NM_001318100.1 | 349 | Missense Mutation | CAC,CGC | H,R 46 | NP_001305029.1 | |
NM_001318101.1 | 349 | Missense Mutation | CAC,CGC | H,R 46 | NP_001305030.1 | |
NM_032429.3 | 349 | Missense Mutation | CAC,CGC | H,R 46 | NP_115805.1 | |
XM_005270222.4 | 349 | Missense Mutation | CAC,CGC | H,R 46 | XP_005270279.1 | |
XM_017016780.1 | 349 | Missense Mutation | CAC,CGC | H,R 46 | XP_016872269.1 | |
XM_017016781.1 | 349 | Missense Mutation | CAC,CGC | H,R 46 | XP_016872270.1 |
PDZD7 - PDZ domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |