Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTTAAAACGGATAAACATGCAC[A/G]GCGATTTCTTTCAAGAACCTTTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611025 MIM: 611260 | ||||||||||||||||||||
Literature Links: |
ENKUR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENKUR - enkurin, TRPC channel interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270383.1 | 418 | Intron | NP_001257312.1 | |||
NM_145010.3 | 418 | Intron | NP_659447.1 | |||
XM_006717389.1 | 418 | Intron | XP_006717452.1 | |||
XM_006717390.1 | 418 | Intron | XP_006717453.1 | |||
XM_011519348.1 | 418 | Intron | XP_011517650.1 | |||
XM_017015830.1 | 418 | Intron | XP_016871319.1 |
THNSL1 - threonine synthase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024838.4 | 418 | Missense Mutation | CAG,CGG | Q,R 30 | NP_079114.3 | |
XM_005252597.2 | 418 | Missense Mutation | CAG,CGG | Q,R 30 | XP_005252654.1 | |
XM_017016665.1 | 418 | Missense Mutation | CAG,CGG | Q,R 30 | XP_016872154.1 |