Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACCTCGAGGGCTGCCTGGCCTG[C/T]GCCACGGCCTACTTCATCTTCCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606276 MIM: 614189 | ||||||||||||||||||||
Literature Links: |
CRTAC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRTAC1 - cartilage acidic protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206528.2 | 309 | Intron | NP_001193457.1 | |||
NM_018058.6 | 309 | Intron | NP_060528.3 | |||
XM_005269938.4 | 309 | Intron | XP_005269995.1 | |||
XM_011539917.1 | 309 | Intron | XP_011538219.1 | |||
XM_017016366.1 | 309 | Intron | XP_016871855.1 | |||
XM_017016367.1 | 309 | Intron | XP_016871856.1 |
GOLGA7B - golgin A7 family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010917.2 | 309 | Silent Mutation | TGC,TGT | C,C 81 | NP_001010917.1 |