Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTCATTAGATTAATAATTTTA[G/T]TATAAAGCCTCCAAGGGGCATCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604551 MIM: 613497 | ||||||||||||||||||||
Literature Links: |
CH25H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CH25H - cholesterol 25-hydroxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIPA - lipase A, lysosomal acid type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000235.3 | 1470 | Missense Mutation | AAT,ACT | N,T 389 | NP_000226.2 | |
NM_001127605.2 | 1470 | Missense Mutation | AAT,ACT | N,T 389 | NP_001121077.1 | |
NM_001288979.1 | 1470 | Missense Mutation | AAT,ACT | N,T 273 | NP_001275908.1 |