Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCGAGAGTGGGGTGGGCAAGTC[C/G]AGGTGAGGCGGAGGTGCGGGTCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602207 | ||||||||||||||||||||
Literature Links: |
RAB18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB18 - RAB18, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256410.1 | 262 | Silent Mutation | TCC,TCG | S,S 22 | NP_001243339.1 | |
NM_001256411.1 | 262 | Silent Mutation | TCC,TCG | S,S 22 | NP_001243340.1 | |
NM_001256412.1 | 262 | Silent Mutation | TCC,TCG | S,S 22 | NP_001243341.1 | |
NM_001256415.1 | 262 | Silent Mutation | TCC,TCG | S,S 22 | NP_001243344.1 | |
NM_021252.4 | 262 | Silent Mutation | TCC,TCG | S,S 22 | NP_067075.1 |