Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTACAATGTGCAGCCGCCAGAT[A/G]GTATCCTCCTATGGAAAAAAATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610366 MIM: 193065 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3M1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP3M1 - adaptor related protein complex 3 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320263.1 | 1852 | UTR 3 | NP_001307192.1 | |||
NM_001320264.1 | 1852 | UTR 3 | NP_001307193.1 | |||
NM_001320265.1 | 1852 | UTR 3 | NP_001307194.1 | |||
NM_012095.5 | 1852 | UTR 3 | NP_036227.1 | |||
NM_207012.3 | 1852 | UTR 3 | NP_996895.1 |
VCL - vinculin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |