Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTTTCTTAAAGAACATCGATA[C/T]ATTGAACTCTTCTTGAATTCTACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602324 MIM: 612189 MIM: 610328 | ||||||||||||||||||||
Literature Links: |
HNRNPH3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPH3 - heterogeneous nuclear ribonucleoprotein H3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322434.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 262 | NP_001309363.1 | |
NM_001322436.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 262 | NP_001309365.1 | |
NM_001322437.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 262 | NP_001309366.1 | |
NM_001322438.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 247 | NP_001309367.1 | |
NM_001322439.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 247 | NP_001309368.1 | |
NM_001322440.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 247 | NP_001309369.1 | |
NM_001322441.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 247 | NP_001309370.1 | |
NM_001322442.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 256 | NP_001309371.1 | |
NM_001322443.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 256 | NP_001309372.1 | |
NM_001322444.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 154 | NP_001309373.1 | |
NM_001322445.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 131 | NP_001309374.1 | |
NM_001322446.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 131 | NP_001309375.1 | |
NM_001322447.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 131 | NP_001309376.1 | |
NM_001322448.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 131 | NP_001309377.1 | |
NM_001322449.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 131 | NP_001309378.1 | |
NM_001322450.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 125 | NP_001309379.1 | |
NM_001322451.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 93 | NP_001309380.1 | |
NM_001322452.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 93 | NP_001309381.1 | |
NM_001322453.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 93 | NP_001309382.1 | |
NM_012207.2 | 997 | Silent Mutation | TAC,TAT | Y,Y 262 | NP_036339.1 | |
NM_021644.3 | 997 | Silent Mutation | TAC,TAT | Y,Y 247 | NP_067676.2 | |
XM_011539742.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 154 | XP_011538044.1 | |
XM_017016168.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 262 | XP_016871657.1 | |
XM_017016169.1 | 997 | Silent Mutation | TAC,TAT | Y,Y 131 | XP_016871658.1 |
PBLD - phenazine biosynthesis like protein domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUFY2 - RUN and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042417.1 | 997 | Intron | NP_001035882.1 | |||
NM_001278225.1 | 997 | Intron | NP_001265154.1 | |||
NM_017987.4 | 997 | Intron | NP_060457.4 | |||
XM_005269953.4 | 997 | Intron | XP_005270010.1 | |||
XM_005269955.4 | 997 | Intron | XP_005270012.1 | |||
XM_005269956.4 | 997 | Intron | XP_005270013.1 | |||
XM_005269957.4 | 997 | Intron | XP_005270014.1 | |||
XM_006717911.3 | 997 | Intron | XP_006717974.1 | |||
XM_011539942.2 | 997 | Intron | XP_011538244.1 |