Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCCCAGGTGGATTCATACGAC[A/G]TGACTGTGGACGAGGAACTGGGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALOX5 - arachidonate 5-lipoxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000698.4 | 280 | Missense Mutation | ATG,GTG | M,V 56 | NP_000689.1 | |
NM_001256153.2 | 280 | Missense Mutation | ATG,GTG | M,V 56 | NP_001243082.1 | |
NM_001256154.2 | 280 | Missense Mutation | ATG,GTG | M,V 56 | NP_001243083.1 | |
NM_001320861.1 | 280 | Missense Mutation | ATG,GTG | M,V 56 | NP_001307790.1 | |
NM_001320862.1 | 280 | UTR 5 | NP_001307791.1 | |||
XM_017016012.1 | 280 | UTR 5 | XP_016871501.1 |